siRNA Details
Gene Symbol: TCF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322151.1 | NP_001309080.1 | 3, 4 | 445 | ||
NM_001322152.1 | NP_001309081.1 | 3, 4 | 450 | |||
NM_001322154.1 | NP_001309083.1 | 3, 4 | 450 | |||
NM_001322156.1 | NP_001309085.1 | 3, 4 | 450 | |||
NM_001322157.1 | NP_001309086.1 | 3, 4 | 450 | |||
NM_001322158.1 | NP_001309087.1 | 3, 4 | 450 | |||
NM_001322159.1 | NP_001309088.1 | 3, 4 | 450 | |||
NM_001322161.1 | NP_001309090.1 | 3, 4 | 296 | |||
NM_001322162.1 | NP_001309091.1 | 3, 4 | 296 | |||
NM_001322165.1 | NP_001309094.1 | 2, 3 | 279 | |||
NM_003205.3 | NP_003196.1 | 3, 4 | 450 | |||
NM_207036.1 | NP_996919.1 | 3, 4 | 440 | |||
NM_207037.1 | NP_996920.1 | 3, 4 | 450 | |||
NM_207038.1 | NP_996921.1 | 3, 4 | 440 | |||
XM_011521965.1 | XP_011520267.1 | 3, 4 | 297 | |||
XM_011521967.1 | XP_011520269.1 | 3, 4 | 297 | |||
XM_017022520.1 | XP_016878009.1 | 3, 4 | 416 | |||
GenBank mRNA | AK302923.1 | - | 3, 4 | 400 | ||
AK312710.1 | - | 3, 4 | 410 | |||
AL831981.2 | - | 3, 4 | 418 | |||
BC050556.1 | - | 3, 4 | 391 | 5767579 | ||
BU178185.1 | - | 3, 4 | 461 | |||
M80627.1 | - | 3, 4 | 369 | |||
M83233.1 | - | 3, 4 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
