siRNA Details
Gene Symbol: TEAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003213.3 | NP_003204.2 | 3 | 488 | ||
NM_201441.2 | NP_958849.1 | 3 | 488 | |||
GenBank mRNA | AK222947.1 | - | 3 | 375 | ||
AY101179.1 | - | 3 | 470 | |||
BC015497.1 | - | 3 | 386 | 3913870 | ||
BM464237.1 | - | 470 | ||||
DA210894.1 | - | 3 | 488 | |||
U63824.1 | - | 3 | 359 | |||
X94438.1 | - | 1 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
