siRNA Details
Gene Symbol: FOXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012182.2 | NP_036314.2 | 2 | 597 | ||
GenBank mRNA | AF071554.1 | - | 1 | 145 | ||
AK290944.1 | - | 2 | 597 | |||
AK314075.1 | - | 2 | 425 | |||
BC113710.1 | - | 1 | 180 | 8322762 | ||
U87393.1 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
