siRNA Details
Gene Symbol: FOXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012182.2 | NP_036314.2 | 2 | 601 | ||
GenBank mRNA | AF071554.1 | - | 1 | 149 | ||
AK290944.1 | - | 2 | 601 | |||
AK314075.1 | - | 2 | 429 | |||
BC113710.1 | - | 1 | 184 | 8322762 | ||
U87393.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
