siRNA Details
Gene Symbol: OR3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012373.2 | NP_036505.2 | 1 | 679 | ||
XM_011524028.2 | XP_011522330.1 | 2 | 689 | |||
GenBank mRNA | BC069415.1 | - | 1 | 679 | 7262255 | |
BC108921.2 | - | 1 | 673 | 40023068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
