siRNA Details
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 15 | 1685 | ||
XM_017015019.1 | XP_016870508.1 | 15 | 1730 | |||
XM_017015020.1 | XP_016870509.1 | 14 | 1685 | |||
XM_017015021.1 | XP_016870510.1 | 14 | 1673 | |||
XM_017015022.1 | XP_016870511.1 | 14 | 1643 | |||
XM_017015023.1 | XP_016870512.1 | 13 | 1628 | |||
XM_017015024.1 | XP_016870513.1 | 13 | 1598 | |||
XM_017015025.1 | XP_016870514.1 | 13 | 1586 | |||
XM_017015026.1 | XP_016870515.1 | 14 | 1600 | |||
XM_017015028.1 | XP_016870517.1 | 16 | 1598 | |||
XM_017015029.1 | XP_016870518.1 | 12 | 1549 | |||
XM_017015031.1 | XP_016870520.1 | 11 | 1241 | |||
XM_017015033.1 | XP_016870522.1 | 10 | 1154 | |||
XM_017015034.1 | XP_016870523.1 | 10 | 1104 | |||
XM_017015035.1 | XP_016870524.1 | 14 | 1688 | |||
XM_017015036.1 | XP_016870525.1 | 13 | 1601 | |||
GenBank mRNA | CR627025.1 | - | 2 | 3421 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 16 | 1783 | ||
NM_001145356.1 | NP_001138828.1 | 14 | 1628 | |||
NM_018491.3 | NP_060961.3 | 15 | 1685 | |||
XM_005251513.4 | XP_005251570.1 | 14 | 1633 | |||
XM_005251514.1 | XP_005251571.1 | 14 | 1598 | |||
XM_005251515.1 | XP_005251572.1 | 13 | 1541 | |||
XM_011517961.2 | XP_011516263.1 | 15 | 1720 | |||
XM_011517963.1 | XP_011516265.1 | 12 | 1223 | |||
GenBank mRNA | AF257330.2 | - | 15 | 1643 | ||
AY343911.1 | - | 15 | 1642 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 15 | 1757 | ||
XM_005263600.2 | XP_005263657.1 | 14 | 1700 | |||
XM_005263602.2 | XP_005263659.1 | 14 | 1670 | |||
XM_005263603.2 | XP_005263660.1 | 16 | 1791 | |||
XM_005263604.2 | XP_005263661.1 | 13 | 1613 | |||
XM_011510667.1 | XP_011508969.1 | 14 | 1627 | |||
XM_011510668.2 | XP_011508970.1 | 12 | 1317 | |||
XM_011510669.2 | XP_011508971.1 | 16 | 1801 | |||
XM_017003408.1 | XP_016858897.1 | 17 | 1921 | |||
GenBank mRNA | AY343910.1 | - | 15 | 1642 | ||
DA296279.1 | - | 1 | 496 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 14 | 1606 | ||
NM_201453.3 | NP_958861.2 | 15 | 1666 | |||
XM_005277637.2 | XP_005277694.1 | 14 | 1688 | |||
XM_005277639.2 | XP_005277696.1 | 14 | 1658 | |||
XM_005277640.2 | XP_005277697.1 | 13 | 1601 | |||
XM_006717116.1 | XP_006717179.1 | 14 | 1615 | |||
XM_011518706.2 | XP_011517008.1 | 10 | 1196 | |||
XM_017014730.1 | XP_016870219.1 | 13 | 1598 | |||
XM_017014731.1 | XP_016870220.1 | 15 | 4117 | |||
XM_017014732.1 | XP_016870221.1 | 9 | 1109 | |||
XM_017014733.1 | XP_016870222.1 | 10 | 6416 | |||
GenBank mRNA | AY344633.1 | - | 15 | 1642 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 13 | 1578 | ||
NM_001286835.1 | NP_001273764.1 | 17 | 2830 | |||
XM_005272748.1 | XP_005272805.1 | 15 | 1682 | |||
XM_005272749.1 | XP_005272806.1 | 14 | 1625 | |||
XM_005272750.1 | XP_005272807.1 | 14 | 1622 | |||
XM_005272751.1 | XP_005272808.1 | 14 | 1595 | |||
XM_006716914.1 | XP_006716977.1 | 14 | 1552 | |||
XM_017014455.1 | XP_016869944.1 | 13 | 1465 | |||
XM_017014456.1 | XP_016869945.1 | 16 | 1613 | |||
XM_017014457.1 | XP_016869946.1 | 15 | 1526 | |||
XM_017014459.1 | XP_016869948.1 | 17 | 1861 | |||
XM_017014461.1 | XP_016869950.1 | 16 | 1774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
