siRNA Details
Gene Symbol: RPL23AP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002229.2 | 1 | 282 | |||
GenBank mRNA | BC096706.1 | - | 1 | 282 | ||
BC096740.1 | - | 1 | 282 | |||
BC098172.1 | - | 1 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
