siRNA Details
Gene Symbol: SLX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024044.3 | NP_076949.1 | 3, 4 | 823 | ||
GenBank mRNA | AK027092.1 | - | 3, 4 | 769 | ||
AK225436.1 | - | 3, 4 | 769 | |||
BC000754.2 | - | 3, 4 | 770 | 2823474 | ||
BC000803.1 | - | 3, 4 | 617 | 3452236 | ||
BC015990.2 | - | 3, 4 | 776 | 3503887 | ||
BC019306.1 | - | 3, 4 | 770 | 2823474 |
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014999.2 | NP_001014999.1 | 3, 4 | 823 | ||
GenBank mRNA | CD673028.1 | - | 3, 4 | 608 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 2, 3 | 700 | |||
GenBank mRNA | AK122707.1 | - | 2, 3 | 700 | ||
AK122733.1 | - | 3, 4 | 528 | |||
BC111011.1 | - | 1, 2 | 728 | 5768190 |
Gene Symbol: SLX1B-SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037609.1 | 2, 3 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
