siRNA Details
Gene Symbol: MGST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260511.1 | NP_001247440.1 | 4 | 472 | ||
NM_001260512.1 | NP_001247441.1 | 5 | 623 | |||
NM_020300.4 | NP_064696.1 | 4 | 479 | |||
NM_145764.2 | NP_665707.1 | 4 | 462 | |||
NM_145791.2 | NP_665734.1 | 4 | 532 | |||
NM_145792.2 | NP_665735.1 | 4 | 454 | |||
NR_048545.1 | 3 | 384 | ||||
NR_048547.1 | 3 | 437 | ||||
GenBank mRNA | AA314967.1 | - | 3 | 437 | ||
AK123482.1 | - | 1 | 1396 | |||
AK291148.1 | - | 4 | 468 | |||
BC005923.1 | - | 4 | 447 | 4297300 | ||
BG569388.1 | - | 4 | 463 | |||
BG617746.1 | - | 4 | 480 | |||
BG778603.1 | - | 3 | 385 | |||
BT006982.1 | - | 3 | 389 | |||
CR407629.1 | - | 3 | 389 | |||
J03746.1 | - | 4 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
