siRNA Details
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014999.2 | NP_001014999.1 | 6 | 1135 | ||
NM_001015000.2 | NP_001015000.1 | 5 | 793 |
Gene Symbol: SLX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024044.3 | NP_076949.1 | 6 | 1135 | ||
NM_178044.2 | NP_835145.1 | 5 | 793 | |||
GenBank mRNA | AK027092.1 | - | 5, 6 | 999 | ||
AK225436.1 | - | 5, 6 | 998 | |||
BC000754.2 | - | 6 | 1082 | 2823474 | ||
BC000803.1 | - | 6 | 929 | 3452236 | ||
BC015990.2 | - | 6 | 1088 | 3503887 | ||
BC019306.1 | - | 6 | 1082 | 2823474 | ||
BC069007.1 | - | 5 | 732 | 4335620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
