siRNA Details
Gene Symbol: FER1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_119376.1 | 35, 36 | 4387 | |||
GenBank mRNA | AK023521.1 | - | 488 | |||
AY461812.1 | - | 35, 36 | 4554 | |||
BX648024.1 | - | 9, 10 | 1210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
