siRNA Details
Gene Symbol: OR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030905.2 | NP_112167.2 | 1 | 340 | ||
GenBank mRNA | BC069121.1 | - | 1 | 238 | 7216975 | |
BC108896.1 | - | 1 | 241 | 40003949 | ||
BC108897.1 | - | 1 | 238 | 40003955 | ||
BC108898.1 | - | 1 | 238 | 40003956 | ||
BC108899.1 | - | 1 | 238 | 40003957 | ||
BU171389.1 | - | 1 | 342 |
Gene Symbol: OR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005216.3 | NP_001005216.2 | 1 | 325 | ||
GenBank mRNA | BC136973.1 | - | 1 | 325 | 9020965 | |
BC136975.1 | - | 1 | 326 | 9020967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
