siRNA Details
Gene Symbol: RXRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291920.1 | NP_001278849.1 | 3, 4 | 1279 | ||
NM_001291921.1 | NP_001278850.1 | 2, 3 | 1070 | |||
NM_002957.5 | NP_002948.1 | 3, 4 | 607 | |||
GenBank mRNA | AB307705.1 | - | 3, 4 | 438 | ||
AK131192.1 | - | 2, 3 | 1070 | |||
AK131513.1 | - | 3, 4 | 1279 | |||
BC063827.1 | - | 3, 4 | 600 | |||
BC110998.1 | - | 3, 4 | 569 | 5723745 | ||
BQ220698.1 | - | 3, 4 | 608 | |||
HQ692843.1 | - | 3, 4 | 438 | |||
X52773.1 | - | 3, 4 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
