siRNA Details
Gene Symbol: SLC5A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042450.2 | NP_001035915.1 | 7, 8 | 689 | ||
NM_001270649.1 | NP_001257578.1 | 7, 8 | 689 | |||
NM_152351.4 | NP_689564.3 | 7, 8 | 689 | |||
XM_011523652.1 | XP_011521954.1 | 7, 8 | 845 | |||
XM_017024191.1 | XP_016879680.1 | 7, 8 | 846 | |||
GenBank mRNA | AK057946.1 | - | 7, 8 | 690 | ||
AK298345.1 | - | 7, 8 | 675 | |||
BC034380.1 | - | 7, 8 | 634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
