siRNA Details
Gene Symbol: GGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152657.3 | NP_689870.3 | 3 | 922 | ||
XM_005258619.4 | XP_005258676.1 | 3 | 1284 | |||
XM_011526603.2 | XP_011524905.1 | 3 | 1034 | |||
XM_017026451.1 | XP_016881940.1 | 2 | 1128 | |||
GenBank mRNA | AF538035.1 | - | 3 | 927 | ||
AK057356.2 | - | 3 | 662 | |||
AK093032.1 | - | 2 | 870 | |||
BC035008.2 | - | 3 | 922 | 5269088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
