siRNA Details
Gene Symbol: SCN8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177984.2 | NP_001171455.1 | 26 | 8967 | ||
NM_014191.3 | NP_055006.1 | 27 | 9090 | |||
XM_006719556.3 | XP_006719619.1 | 27 | 9681 | |||
XM_011538650.2 | XP_011536952.1 | 27 | 9109 | |||
XM_011538651.2 | XP_011536953.1 | 27 | 9074 | |||
XM_017019794.1 | XP_016875283.1 | 27 | 9087 | |||
XM_017019795.1 | XP_016875284.1 | 26 | 8983 | |||
GenBank mRNA | AK091315.1 | - | 1 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
