siRNA Details
Gene Symbol: FAM27E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103833.1 | 2 | 367 | |||
GenBank mRNA | BC032035.1 | - | 1 | 201 | 4826036 | |
BC044751.1 | - | 1 | 201 | |||
BC119675.1 | - | 2 | 366 | 40116244 | ||
BC148261.1 | - | 1 | 542 |
Gene Symbol: FAM27E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103714.2 | 1 | 545 | |||
GenBank mRNA | BC119676.1 | - | 1 | 546 |
Gene Symbol: LOC107987001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015381.1 | XP_016870870.1 | 2 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
