siRNA Details
Gene Symbol: SHISA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173461.1 | NP_001166932.1 | 4 | 5562 | ||
NM_001173462.1 | NP_001166933.1 | 5 | 5658 | |||
NM_207386.3 | NP_997269.2 | 6 | 5715 | |||
XM_017024618.1 | XP_016880107.1 | 5 | 5619 | |||
GenBank mRNA | AK094349.1 | - | 1 | 607 | ||
AK095052.1 | - | 1 | 1208 | |||
BC105704.1 | - | 1 | 364 | |||
BC105705.1 | - | 1 | 364 | |||
BC105706.1 | - | 1 | 363 | |||
BC105707.1 | - | 1 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
