siRNA Details
Gene Symbol: FHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039492.2 | NP_001034581.1 | 4 | 442 | ||
NM_001318894.1 | NP_001305823.1 | 3 | 688 | |||
NM_001318895.1 | NP_001305824.1 | 4 | 676 | |||
NM_001318896.1 | NP_001305825.1 | 3 | 391 | |||
NM_001318899.1 | NP_001305828.1 | 5 | 564 | |||
NM_001450.3 | NP_001441.4 | 3 | 625 | |||
NM_201555.1 | NP_963849.1 | 5 | 797 | |||
NM_201557.3 | NP_963851.2 | 3 | 617 | |||
XM_011510798.2 | XP_011509100.1 | 4 | 424 | |||
GenBank mRNA | AK313841.1 | - | 4 | 474 | ||
AL521653.3 | - | 3 | 318 | |||
AY634685.1 | - | 2 | 218 | |||
BC012742.2 | - | 5 | 797 | 3840517 | ||
BC014397.1 | - | 5 | 797 | 3546686 | ||
BC093049.1 | - | 3 | 383 | 30374919 | ||
BM559077.1 | - | 5 | 446 | |||
BT006960.1 | - | 2 | 218 | |||
CF552311.1 | - | 3 | 348 | |||
CR457437.1 | - | 2 | 218 | |||
CR936682.1 | - | 3 | 688 | |||
CR936799.1 | - | 3 | 393 | |||
DQ266432.1 | - | 4 | 676 | |||
DQ307067.1 | - | 3 | 555 | |||
JQ313902.1 | - | 2 | 188 | |||
L42176.1 | - | 4 | 356 | |||
U29332.1 | - | 3 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
