siRNA Details
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267810.1 | NP_001254739.1 | 4 | 302 | ||
NM_002212.3 | NP_002203.1 | 4 | 372 | |||
NM_181468.2 | NP_852133.1 | 3 | 506 | |||
NR_052023.1 | 4 | 365 | ||||
GenBank mRNA | AB062289.1 | - | 4 | 305 | ||
AF022229.1 | - | 3 | 361 | |||
AF047433.1 | - | 4 | 346 | |||
AK314969.1 | - | 4 | 348 | |||
AL537456.3 | - | 3 | 499 | |||
BC001119.2 | - | 4 | 345 | 2989973 | ||
BC011845.2 | - | 4 | 323 | 4120463 | ||
BC019305.1 | - | 4 | 305 | 2823211 | ||
BQ222221.1 | - | 3 | 292 | |||
CN265282.1 | - | 3 | 506 | |||
CR456764.1 | - | 3 | 276 | |||
CT002623.1 | - | 4 | 317 | |||
DB448068.1 | - | 4 | 373 | |||
Y11435.1 | - | 4 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
