siRNA Details
Gene Symbol: CCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002984.3 | NP_002975.1 | 2, 3 | 267 | ||
GenBank mRNA | AY766446.1 | - | 2, 3 | 267 | ||
BC104226.1 | - | 2, 3 | 258 | 40032168 | ||
BC104227.1 | - | 2, 3 | 258 | 40032172 | ||
BC107433.1 | - | 2, 3 | 268 | 6727378 | ||
CR542119.1 | - | 2, 3 | 188 | |||
J04130.1 | - | 2, 3 | 296 | |||
M23502.1 | - | 2, 3 | 262 | |||
M25316.1 | - | 2, 3 | 261 | |||
M57503.1 | - | 2, 3 | 175 | |||
X16166.1 | - | 2, 3 | 240 | |||
X53683.1 | - | 2, 3 | 254 |
Gene Symbol: CCL4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207007.3 | NP_996890.1 | 2, 3 | 422 | ||
GenBank mRNA | AY766447.1 | - | 2, 3 | 267 | ||
BC070310.1 | - | 2, 3 | 264 | 6708338 | ||
BC130456.1 | - | 2, 3 | 229 | 40146487 | ||
BC130458.1 | - | 2, 3 | 229 | 40146489 | ||
BC144393.1 | - | 2, 3 | 229 | 9052914 | ||
BC144394.1 | - | 2, 3 | 229 | 9052915 | ||
BC146944.1 | - | 2, 3 | 422 | 9056872 | ||
BC146958.1 | - | 2, 3 | 422 | 9056886 | ||
BC171857.1 | - | 2, 3 | 422 | 9054511 | ||
BC171864.1 | - | 2, 3 | 422 | 9054518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
