siRNA Details
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308078.1 | NP_001295007.1 | 3 | 274 | ||
GenBank mRNA | AK307158.1 | - | 2 | 657 | ||
AK310053.1 | - | 3 | 277 | |||
BC009401.2 | - | 3 | 411 | 4300407 | ||
BF568456.1 | - | 3 | 412 | |||
M59807.1 | - | 3 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
