siRNA Details
Gene Symbol: GPM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001994.2 | NP_001001994.1 | 7 | 1027 | ||
NM_001001996.2 | NP_001001996.1 | 8 | 1345 | |||
NM_005278.4 | NP_005269.1 | 7 | 1225 | |||
XM_017029432.1 | XP_016884921.1 | 8 | 1035 | |||
XM_017029434.1 | XP_016884923.1 | 7 | 752 | |||
GenBank mRNA | AK294314.1 | - | 7 | 798 | ||
BC008151.1 | - | 7 | 851 | 4215416 | ||
BC047295.1 | - | 8 | 1055 | 4215095 | ||
U45955.1 | - | 7 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
