siRNA Details
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006927.3 | NP_008858.1 | 2 | 1411 | ||
GenBank mRNA | AK127322.1 | - | 1 | 2376 | ||
AK313020.1 | - | 1 | 542 | |||
BC036777.1 | - | 1 | 310 | 5750330 | ||
U63090.1 | - | 1 | 948 | |||
X96667.1 | - | 1 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
