siRNA Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014451.2 | NP_001014451.1 | 2, 3 | 244 | ||
NM_001257172.1 | NP_001244101.1 | 2, 3 | 169 | |||
NM_007242.5 | NP_009173.1 | 2, 3 | 244 | |||
XM_011522832.2 | XP_011521134.1 | 3, 4 | 300 | |||
XM_011522833.2 | XP_011521135.1 | 3, 4 | 300 | |||
XM_011522834.2 | XP_011521136.1 | 2, 3 | 189 | |||
XM_017022890.1 | XP_016878379.1 | 2, 3 | 282 | |||
GenBank mRNA | AJ237946.1 | - | 2, 3 | 105 | ||
AK027378.1 | - | 2, 3 | 209 | |||
AK223115.1 | - | 2, 3 | 215 | |||
AK302107.1 | - | 2, 3 | 169 | |||
AL136639.1 | - | 2, 3 | 213 | |||
BC003626.2 | - | 2, 3 | 208 | 3030038 | ||
BC110805.1 | - | 1, 2 | 15 | |||
CR457215.1 | - | 2, 3 | 105 | |||
CR533509.1 | - | 2, 3 | 105 | |||
DC385044.1 | - | 2, 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
