siRNA Details
Gene Symbol: AKR7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012067.2 | NP_036199.2 | 1, 2 | 297 | ||
XM_011541046.2 | XP_011539348.1 | 1, 2 | 873 | |||
GenBank mRNA | AF040639.1 | - | 1, 2 | 304 | ||
AJ271799.1 | - | 1, 2 | 227 | |||
BC025709.1 | - | 1, 2 | 283 | 5224626 | ||
BC031562.1 | - | 1, 2 | 239 | 5184829 | ||
BC042420.1 | - | 1, 2 | 263 | 5186889 | ||
BM855386.1 | - | 1, 2 | 309 | |||
BM920315.1 | - | 1, 2 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
