siRNA Details
Gene Symbol: POFUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015352.1 | NP_056167.1 | 2 | 234 | ||
NM_172236.1 | NP_758436.1 | 2 | 234 | |||
GenBank mRNA | AF375884.1 | - | 2 | 234 | ||
AK291033.1 | - | 2 | 241 | |||
AL832023.1 | - | 2 | 166 | |||
BC000582.2 | - | 2 | 234 | 3162533 | ||
D80002.2 | - | 2 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
