siRNA Details
Gene Symbol: ZNF444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253792.1 | NP_001240721.1 | 4 | 785 | ||
XM_011527073.1 | XP_011525375.1 | 5 | 2434 | |||
GenBank mRNA | AB052954.1 | - | 4 | 628 | ||
AK001999.1 | - | 4 | 630 | |||
BC021282.2 | - | 4 | 615 | 5016494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
