siRNA Details
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 7 | 983 | ||
XM_005263396.3 | XP_005263453.1 | 8 | 1408 | |||
XM_005263397.1 | XP_005263454.1 | 5 | 509 | |||
XM_011518272.1 | XP_011516574.1 | 7 | 677 | |||
XM_011518273.2 | XP_011516575.1 | 6 | 508 | |||
XM_011518274.1 | XP_011516576.1 | 5 | 538 | |||
XM_017014301.1 | XP_016869790.1 | 7 | 1264 | |||
XM_017014302.1 | XP_016869791.1 | 7 | 1188 | |||
GenBank mRNA | AF116701.2 | - | 7 | 962 | ||
AJ519841.1 | - | 5 | 508 | |||
BC130580.1 | - | 7 | 642 | 40146611 | ||
BC144082.1 | - | 7 | 642 | 9052602 | ||
BU949503.1 | - | 5 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
