siRNA Details
Gene Symbol: TNRC6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142640.1 | NP_001136112.1 | 22 | 7211 | ||
NM_018996.3 | NP_061869.2 | 21 | 7103 | |||
XM_006721996.3 | XP_006722059.3 | 23 | 12872 | |||
XM_006721997.3 | XP_006722060.3 | 22 | 12722 | |||
XM_006722000.3 | XP_006722063.3 | 22 | 12746 | |||
XM_017024906.1 | XP_016880395.1 | 23 | 12863 | |||
XM_017024907.1 | XP_016880396.1 | 22 | 12755 | |||
XM_017024908.1 | XP_016880397.1 | 22 | 12647 | |||
GenBank mRNA | AK000022.1 | - | 1 | 832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
