siRNA Details
Gene Symbol: NR2C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291694.1 | NP_001278623.1 | 3, 4 | 636 | ||
NM_003298.4 | NP_003289.2 | 4, 5 | 693 | |||
XM_011534058.2 | XP_011532360.1 | 7, 8 | 728 | |||
XM_011534059.2 | XP_011532361.1 | 7, 8 | 1634 | |||
XM_011534061.2 | XP_011532363.1 | 5, 6 | 582 | |||
XM_011534063.2 | XP_011532365.1 | 6, 7 | 669 | |||
XM_011534064.2 | XP_011532366.1 | 6, 7 | 831 | |||
XM_011534065.2 | XP_011532367.1 | 5, 6 | 541 | |||
XM_011534066.2 | XP_011532368.1 | 4, 5 | 1344 | |||
XM_017007118.1 | XP_016862607.1 | 5, 6 | 810 | |||
XM_017007119.1 | XP_016862608.1 | 5, 6 | 470 | |||
XM_017007120.1 | XP_016862609.1 | 3, 4 | 478 | |||
GenBank mRNA | AB307708.1 | - | 2, 3 | 274 | ||
AK290590.1 | - | 3, 4 | 636 | |||
BC051670.1 | - | 4, 5 | 466 | 5272493 | ||
BF307171.1 | - | 3, 4 | 292 | |||
HQ995528.1 | - | 3, 4 | 331 | |||
L27586.1 | - | 4, 5 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
