siRNA Details
Gene Symbol: C6orf106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024294.3 | NP_077270.1 | 3 | 616 | ||
XM_005249298.2 | XP_005249355.1 | 3 | 563 | |||
GenBank mRNA | AK313410.1 | - | 3 | 522 | ||
BC002328.1 | - | 3 | 530 | 3504258 | ||
BC010184.1 | - | 3 | 530 | 3959622 | ||
BC075810.1 | - | 3 | 538 | 5535440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
