siRNA Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719532.1 | XP_006719595.1 | 7 | 1251 | ||
XM_006719534.1 | XP_006719597.1 | 7 | 1406 | |||
XM_006719535.3 | XP_006719598.1 | 8 | 1334 | |||
XM_006719536.2 | XP_006719599.1 | 7 | 1223 | |||
XM_011538622.1 | XP_011536924.1 | 8 | 1333 | |||
XM_011538623.1 | XP_011536925.1 | 8 | 1520 | |||
XM_017019726.1 | XP_016875215.1 | 7 | 1819 | |||
XM_017019727.1 | XP_016875216.1 | 7 | 1619 | |||
XM_017019728.1 | XP_016875217.1 | 7 | 1610 | |||
XM_017019729.1 | XP_016875218.1 | 7 | 1819 | |||
XM_017019730.1 | XP_016875219.1 | 7 | 1819 | |||
XM_017019731.1 | XP_016875220.1 | 8 | 1704 | |||
XM_017019732.1 | XP_016875221.1 | 7 | 1819 | |||
XM_017019733.1 | XP_016875222.1 | 7 | 1819 | |||
XM_017019734.1 | XP_016875223.1 | 7 | 1131 | |||
XM_017019735.1 | XP_016875224.1 | 7 | 1819 | |||
GenBank mRNA | AB209034.1 | - | 7 | 1229 | ||
AK026695.1 | - | 1 | 143 | |||
BC052611.1 | - | 1 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
