siRNA Details
Gene Symbol: USF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003367.3 | NP_003358.1 | 5, 6 | 712 | ||
NM_207291.2 | NP_997174.1 | 4, 5 | 511 | |||
XM_005259197.3 | XP_005259254.1 | 5, 6 | 611 | |||
XM_011527260.2 | XP_011525562.1 | 5, 6 | 611 | |||
XM_011527261.2 | XP_011525563.1 | 5, 6 | 611 | |||
XM_017027199.1 | XP_016882688.1 | 4, 5 | 410 | |||
GenBank mRNA | AK297022.1 | - | 5, 6 | 582 | ||
BC049821.1 | - | 5, 6 | 625 | 5483523 | ||
CR536504.1 | - | 5, 6 | 588 | |||
M77476.1 | - | 2, 3 | 312 | |||
S50537.1 | - | 1, 2 | 252 | |||
X90824.1 | - | 3, 4 | 451 | |||
X90825.1 | - | 4, 5 | 496 | |||
X90826.1 | - | 5, 6 | 594 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
