siRNA Details
Gene Symbol: OR7A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030901.1 | NP_112163.1 | 1 | 216 | ||
GenBank mRNA | BC101587.1 | - | 1 | 313 | 8069093 | |
BC117357.1 | - | 1 | 313 | 40125908 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR7A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030901.1 | NP_112163.1 | 1 | 216 | ||
GenBank mRNA | BC101587.1 | - | 1 | 313 | 8069093 | |
BC117357.1 | - | 1 | 313 | 40125908 |