siRNA Details
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 13 | 3876 | |||
GenBank mRNA | DQ786236.1 | - | 8 | 1013 |
Gene Symbol: OR2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030908.2 | NP_112170.1 | 1 | 305 | ||
GenBank mRNA | BC120953.2 | - | 1 | 305 | 40115181 | |
BC120954.1 | - | 1 | 305 | 40115185 |
Gene Symbol: OR2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005328.1 | NP_001005328.1 | 1 | 305 | ||
GenBank mRNA | BC136704.1 | - | 1 | 399 | 9020694 | |
BC136711.1 | - | 1 | 399 | 9020701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
