siRNA Details
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046310.1 | 3 | 544 | |||
NR_046435.1 | 3 | 379 | ||||
GenBank mRNA | BC007377.2 | - | 3 | 373 | ||
BY994354.1 | - | 3 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
