siRNA Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 2 | 578 | ||
NM_181268.2 | NP_851785.2 | 2 | 578 | |||
NM_181269.2 | NP_851786.1 | 2 | 283 | |||
NM_181270.2 | NP_851787.1 | 2 | 283 | |||
GenBank mRNA | AF278576.1 | - | 2 | 198 | ||
AF278577.1 | - | 2 | 198 | |||
AY174118.1 | - | 2 | 287 | |||
AY174119.1 | - | 2 | 287 | |||
AY174125.1 | - | 2 | 287 | |||
AY174126.1 | - | 2 | 287 | |||
AY174134.1 | - | 2 | 278 | |||
AY174135.1 | - | 2 | 278 | |||
AY174136.1 | - | 2 | 278 | |||
AY174137.1 | - | 2 | 278 | |||
AY174138.1 | - | 2 | 278 | |||
BC057852.1 | - | 2 | 578 | 5273108 | ||
DQ309766.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
