siRNA Details
Gene Symbol: TSSK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053006.4 | NP_443732.3 | 1 | 705 | ||
GenBank mRNA | AF362953.1 | - | 1 | 185 | ||
BC037781.2 | - | 1 | 236 | 5273397 | ||
CR456587.1 | - | 1 | 127 | |||
L77564.1 | - | 1 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
