siRNA Details
Gene Symbol: C9orf116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048265.1 | NP_001041730.1 | 3 | 384 | ||
NM_144654.2 | NP_653255.1 | 3 | 338 | |||
GenBank mRNA | AK024007.1 | - | 3 | 941 | ||
AY927868.1 | - | 3 | 320 | |||
BC021261.1 | - | 4 | 444 | 4136397 | ||
DR003074.1 | - | 3 | 468 |
