siRNA Details
Gene Symbol: SEPT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321496.1 | NP_001308425.1 | 2 | 466 | ||
NM_001321498.1 | NP_001308427.1 | 2 | 466 | |||
NM_001321499.1 | NP_001308428.1 | 2 | 466 | |||
NM_001321501.1 | NP_001308430.1 | 2 | 466 | |||
NM_001321507.1 | NP_001308436.1 | 2 | 466 | |||
NM_001321509.1 | NP_001308438.1 | 2 | 466 | |||
NM_001321510.1 | NP_001308439.1 | 2 | 466 | |||
NM_001321512.1 | NP_001308441.1 | 2 | 466 | |||
NM_001321513.1 | NP_001308442.1 | 2 | 603 | |||
NM_001321514.1 | NP_001308443.1 | 2 | 294 | |||
NM_144710.4 | NP_653311.1 | 2 | 466 | |||
XM_006712317.1 | XP_006712380.1 | 2 | 466 | |||
XM_011510698.2 | XP_011509000.1 | 2 | 299 | |||
XM_011510700.2 | XP_011509002.1 | 2 | 465 | |||
XM_011510701.2 | XP_011509003.1 | 2 | 507 | |||
XM_011510702.2 | XP_011509004.1 | 2 | 465 | |||
XM_011510703.2 | XP_011509005.1 | 2 | 76 | |||
XM_011510704.2 | XP_011509006.1 | 2 | 466 | |||
GenBank mRNA | AF146760.1 | - | 2 | 213 | ||
AK021681.1 | - | 2 | 466 | |||
AK292038.1 | - | 2 | 184 | |||
AK294427.1 | - | 2 | 353 | |||
AK295560.1 | - | 2 | 176 | |||
BC020502.1 | - | 2 | 160 | 3913356 | ||
BC050345.1 | - | 2 | 403 | 4830029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
