siRNA Details
Gene Symbol: LRRC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291006.1 | NP_001277935.1 | 4 | 631 | ||
NR_027006.1 | 3 | 1565 | ||||
NR_110991.1 | 3 | 1565 | ||||
XM_011530173.2 | XP_011528475.1 | 4 | 610 | |||
XM_011530174.1 | XP_011528476.1 | 4 | 610 | |||
XM_011530175.1 | XP_011528477.1 | 4 | 610 | |||
XM_011530176.2 | XP_011528478.1 | 4 | 610 | |||
XM_011530177.1 | XP_011528479.1 | 4 | 610 | |||
XM_017028797.1 | XP_016884286.1 | 4 | 610 | |||
GenBank mRNA | AK128646.1 | - | 3 | 1544 | ||
BC140948.1 | - | 3 | 1563 | |||
BC140950.1 | - | 3 | 1563 |
Gene Symbol: TUBA3FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003608.1 | 2 | 695 | |||
GenBank mRNA | AK057257.1 | - | 2 | 647 | ||
AK315686.1 | - | 2 | 643 | |||
BC009991.2 | - | 2 | 553 | |||
BI668929.1 | - | 2 | 700 | |||
CN368433.1 | - | 2 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
