siRNA Details
Gene Symbol: SLC47A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024224.1 | XP_016879713.1 | 13, 14 | 1362 | ||
GenBank mRNA | AK055758.1 | - | 12, 13 | 1160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
