siRNA Details
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 7 | 1048 | ||
NM_153449.3 | NP_703150.1 | 3 | 330 | |||
GenBank mRNA | AF481879.1 | - | 3 | 190 | ||
AK126026.1 | - | 7 | 830 | |||
DB069339.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
