siRNA Details
Gene Symbol: MRPL54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172251.2 | NP_758455.1 | 2 | 243 | ||
GenBank mRNA | AA340000.1 | - | 2 | 243 | ||
BC065273.1 | - | 2 | 236 | 6165136 | ||
BF346319.1 | - | 2 | 236 | |||
BM792364.1 | - | 2 | 203 | |||
BQ061807.1 | - | 2 | 231 |
