siRNA Details
Gene Symbol: FBXL19-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024348.1 | 1 | 616 | |||
GenBank mRNA | AK092263.1 | - | 1 | 616 | ||
BC101464.1 | - | 1 | 542 | |||
BC101988.1 | - | 1 | 542 | |||
BC105687.1 | - | 1 | 542 | |||
BC105926.1 | - | 1 | 542 |
