siRNA Details
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021584.2 | NP_067595.1 | 3 | 308 | ||
GenBank mRNA | AF030089.2 | - | 3 | 294 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111053, NM_001195540, AK147831, BC021354, AK164993, and others.
Gene Ontology Categories:
