siRNA Details
Gene Symbol: Glrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022278.1 | NP_071614.1 | 1 | 119 | ||
GenBank mRNA | AF167981.3 | - | 1 | 119 | ||
AF319950.1 | - | 1 | 88 | |||
BC061555.1 | - | 1 | 148 | 6889009 | ||
FQ224532.1 | - | 1 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
