siRNA Details
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030985.4 | NP_112247.2 | 3 | 379 | ||
XM_006253882.2 | XP_006253944.1 | 3 | 418 | |||
XM_008771593.1 | XP_008769815.1 | 2 | 335 | |||
XM_008771594.1 | XP_008769816.1 | 2 | 351 | |||
GenBank mRNA | BC078810.1 | - | 3 | 379 | 7126918 | |
M74054.1 | - | 3 | 374 |
Gene Symbol: Agtr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031009.2 | NP_112271.2 | 2 | 280 | ||
XM_006232186.2 | XP_006232248.1 | 2 | 1450 | |||
XM_006232187.2 | XP_006232249.1 | 2 | 1364 | |||
XM_008760878.1 | XP_008759100.1 | 2 | 2024 | |||
XM_008760879.1 | XP_008759101.1 | 3 | 462 | |||
GenBank mRNA | M90065.1 | - | 2 | 311 | ||
X64052.1 | - | 2 | 268 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
