siRNA Details
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021584.2 | NP_067595.1 | 3 | 314 | ||
GenBank mRNA | AF030089.2 | - | 3 | 300 | ||
AF045469.1 | - | 3 | 199 |

More Information
Other Species Targeted

Gene Ontology Categories:
